siRNA Details
Gene Symbol: OTOGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173591.3 | NP_775862.3 | 15 | 1446 | ||
XM_005268802.3 | XP_005268859.1 | 19 | 2247 | |||
XM_011538191.1 | XP_011536493.1 | 16 | 1523 | |||
XM_011538192.2 | XP_011536494.1 | 13 | 1464 | |||
XM_011538193.1 | XP_011536495.1 | 13 | 1383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
