siRNA Details
Gene Symbol: ANO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012302.2 | NP_001012302.2 | 13, 14 | 1274 | ||
XM_011520048.1 | XP_011518350.1 | 13, 14 | 1274 | |||
XM_011520051.2 | XP_011518353.1 | 13, 14 | 1274 | |||
XM_011520052.2 | XP_011518354.1 | 13, 14 | 1274 | |||
XM_011520053.2 | XP_011518355.1 | 13, 14 | 1370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
