siRNA Details
Gene Symbol: KIAA1656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046312.1 | 2 | 1894 | |||
GenBank mRNA | AB051443.1 | - | 1 | 1259 | ||
CR456347.1 | - | 1 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
