siRNA Details
Gene Symbol: TMEM210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282477.1 | NP_001269406.1 | 2 | 126 | ||
GenBank mRNA | DB459119.1 | - | 2 | 184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
