siRNA Details
Gene Symbol: SPATA45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024601.2 | NP_001019772.1 | 2 | 192 | ||
GenBank mRNA | AY640234.1 | - | 2 | 149 | ||
BC035742.1 | - | 2 | 192 | 5744157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
