siRNA Details
Gene Symbol: SATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012980.2 | NP_001012998.2 | 1 | 199 | ||
XM_011530940.2 | XP_011529242.1 | 2 | 633 | |||
GenBank mRNA | BC043215.1 | - | 1 | 400 | 5295273 | |
BC126401.1 | - | 1 | 385 | 8992117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
