siRNA Details
Gene Symbol: SATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012980.2 | NP_001012998.2 | 5 | 2085 | ||
GenBank mRNA | BC043215.1 | - | 5 | 1560 | 5295273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
