siRNA Details
Gene Symbol: MILR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085423.1 | NP_001078892.1 | 3 | 174 | ||
NM_001291316.1 | NP_001278245.1 | 3 | 174 | |||
XM_017024486.1 | XP_016879975.1 | 3 | 267 | |||
GenBank mRNA | AB542950.1 | - | 3 | 126 | ||
AB542951.1 | - | 3 | 120 | |||
BC018972.1 | - | 3 | 159 | |||
BC053534.1 | - | 2 | 68 | |||
BC130406.1 | - | 3 | 149 | 40146437 | ||
BC136594.1 | - | 3 | 149 | 9020584 | ||
BC144272.1 | - | 3 | 149 | 9052792 | ||
CD687595.1 | - | 3 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
