siRNA Details
Gene Symbol: MILR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085423.1 | NP_001078892.1 | 5 | 747 | ||
NM_001291316.1 | NP_001278245.1 | 4 | 462 | |||
NM_001291317.1 | NP_001278246.1 | 4 | 477 | |||
XM_017024486.1 | XP_016879975.1 | 5 | 840 | |||
XM_017024487.1 | XP_016879976.1 | 5 | 760 | |||
GenBank mRNA | AB542950.1 | - | 5 | 699 | ||
AB542951.1 | - | 4 | 408 | |||
AB542952.1 | - | 4 | 423 | |||
BC053534.1 | - | 4 | 641 | |||
BC130406.1 | - | 5 | 722 | 40146437 | ||
BC136594.1 | - | 5 | 722 | 9020584 | ||
BC144272.1 | - | 4 | 437 | 9052792 | ||
BE296352.1 | - | 2 | 248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
