siRNA Details
Gene Symbol: MILR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085423.1 | NP_001078892.1 | 4 | 567 | ||
NM_001291317.1 | NP_001278246.1 | 3 | 297 | |||
XM_017024486.1 | XP_016879975.1 | 4 | 660 | |||
XM_017024487.1 | XP_016879976.1 | 4 | 580 | |||
GenBank mRNA | AB542950.1 | - | 4 | 519 | ||
AB542952.1 | - | 3 | 243 | |||
BC018972.1 | - | 4 | 552 | |||
BC053534.1 | - | 3 | 461 | |||
BC130406.1 | - | 4 | 542 | 40146437 | ||
BC136594.1 | - | 4 | 542 | 9020584 | ||
BE296352.1 | - | 1 | 68 | |||
CD687595.1 | - | 4 | 582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
