siRNA Details
Gene Symbol: DNHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144666.2 | NP_653267.2 | 14 | 3166 | ||
GenBank mRNA | BG676072.1 | - | 4 | 563 | ||
BX647806.1 | - | 3 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
