siRNA Details
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 20 | 6581 | ||
NM_153050.2 | NP_694690.1 | 20 | 6497 | |||
NM_153051.2 | NP_694691.1 | 19 | 6470 | |||
XM_005261803.2 | XP_005261860.1 | 22 | 6726 | |||
XM_005261804.3 | XP_005261861.1 | 21 | 6643 | |||
XM_005261805.2 | XP_005261862.1 | 21 | 6699 | |||
XM_005261806.2 | XP_005261863.1 | 21 | 6608 | |||
XM_017029028.1 | XP_016884517.1 | 20 | 6500 | |||
XM_017029029.1 | XP_016884518.1 | 19 | 6463 | |||
XM_017029030.1 | XP_016884519.1 | 19 | 6395 | |||
XM_017029031.1 | XP_016884520.1 | 18 | 6356 | |||
XM_017029033.1 | XP_016884522.1 | 15 | 5927 | |||
XM_017029034.1 | XP_016884523.1 | 21 | 7930 | |||
GenBank mRNA | AK056895.1 | - | 1 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
