siRNA Details
Gene Symbol: ELFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128636.2 | NP_001122108.1 | 2 | 1351 | ||
XM_006715725.3 | XP_006715788.1 | 3 | 1518 | |||
XM_006715726.2 | XP_006715789.1 | 3 | 1465 | |||
XM_006715727.3 | XP_006715790.1 | 3 | 1764 | |||
XM_011515397.2 | XP_011513699.1 | 3 | 1556 | |||
XM_011515398.2 | XP_011513700.1 | 4 | 1701 | |||
XM_011515401.2 | XP_011513703.1 | 2 | 4803 | |||
XM_017012203.1 | XP_016867692.1 | 3 | 3059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
