siRNA Details
Gene Symbol: PABPC4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114734.1 | NP_001108206.2 | 2 | 587 | ||
XM_017007727.1 | XP_016863216.1 | 2 | 626 | |||
GenBank mRNA | DR005318.1 | - | 2 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
