siRNA Details
Gene Symbol: DCAF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029955.3 | NP_001025126.2 | 1 | 1474 | ||
GenBank mRNA | AK122914.1 | - | 1 | 1392 | ||
BC035027.1 | - | 1 | 1347 | 4828469 | ||
BC052943.1 | - | 1 | 1414 | 5268527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
