siRNA Details
Gene Symbol: SLC22A31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242757.1 | NP_001229686.1 | 5 | 971 | ||
XM_011522897.2 | XP_011521199.1 | 6 | 1019 | |||
XM_011522899.2 | XP_011521201.1 | 6 | 956 | |||
XM_017022962.1 | XP_016878451.1 | 3 | 621 | |||
XM_017022963.1 | XP_016878452.1 | 3 | 2369 | |||
XM_017022964.1 | XP_016878453.1 | 4 | 1525 | |||
XM_017022965.1 | XP_016878454.1 | 3 | 2310 | |||
XM_017022966.1 | XP_016878455.1 | 4 | 2148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
