siRNA Details
Gene Symbol: AHNAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138420.2 | NP_612429.2 | 7 | 16104 | ||
XM_005267299.1 | XP_005267356.1 | 7 | 15957 | |||
GenBank mRNA | AB095939.1 | - | 1 | 7004 | ||
AK126472.1 | - | 1 | 2586 | |||
AL834367.1 | - | 1 | 1148 | |||
BC049216.1 | - | 1 | 146 | |||
BC090889.1 | - | 3 | 1222 | |||
BC144467.1 | - | 1 | 1488 | |||
BX537923.1 | - | 1 | 1986 | |||
BX538040.1 | - | 1 | 1760 | |||
BX538106.1 | - | 1 | 2226 | |||
BX538133.1 | - | 1 | 1647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
