siRNA Details
Gene Symbol: AHNAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138420.2 | NP_612429.2 | 7 | 16651 | ||
XM_005267299.1 | XP_005267356.1 | 7 | 16504 | |||
GenBank mRNA | AB095939.1 | - | 1 | 7551 | ||
AK095704.1 | - | 1 | 451 | |||
AK126472.1 | - | 1 | 3133 | |||
AL834367.1 | - | 1 | 1695 | |||
BC011859.2 | - | 1 | 353 | |||
BC033372.1 | - | 1 | 259 | |||
BC049216.1 | - | 1 | 693 | |||
BC090889.1 | - | 3 | 1769 | |||
BC144467.1 | - | 1 | 2035 | |||
BX537923.1 | - | 1 | 2533 | |||
BX538040.1 | - | 1 | 2307 | |||
BX538106.1 | - | 1 | 2773 | |||
BX538133.1 | - | 1 | 2194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
