siRNA Details
Gene Symbol: LOC728024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003671.2 | 1 | 264 | |||
GenBank mRNA | BI758198.1 | - | 1 | 268 | ||
CR977513.1 | - | 1 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
