siRNA Details
Gene Symbol: CCDC172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198515.2 | NP_940917.1 | 4, 5 | 535 | ||
XM_006717820.3 | XP_006717883.1 | 4, 5 | 830 | |||
XM_011539771.2 | XP_011538073.1 | 4, 5 | 863 | |||
XM_017016206.1 | XP_016871695.1 | 4, 5 | 453 | |||
GenBank mRNA | AJ457798.1 | - | 3, 4 | 517 | ||
BC044830.1 | - | 4, 5 | 535 | 5166167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
