siRNA Details
Gene Symbol: IGLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF710786.1 | - | 103 | |||
EF710787.1 | - | 103 | ||||
EF710788.1 | - | 103 | ||||
EF710790.1 | - | 103 | ||||
EF710791.1 | - | 103 |
Gene Symbol: IGLV6-57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF267875.1 | - | 1 | 103 | ||
BC023973.1 | - | 2 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
