siRNA Details
Gene Symbol: TMEM120B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080825.2 | NP_001074294.2 | 12 | 5811 | ||
GenBank mRNA | BC032529.1 | - | 1 | 561 | ||
BX648084.1 | - | 8 | 5284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
