siRNA Details
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020759.2 | NP_065810.2 | 9, 10 | 767 | ||
XM_011521831.2 | XP_011520133.1 | 9, 10 | 780 | |||
XM_011521832.2 | XP_011520134.1 | 9, 10 | 777 | |||
XM_011521833.2 | XP_011520135.1 | 8, 9 | 747 | |||
XM_011521834.2 | XP_011520136.1 | 9, 10 | 780 | |||
XM_011521835.2 | XP_011520137.1 | 9, 10 | 780 | |||
XM_011521836.2 | XP_011520138.1 | 9, 10 | 1082 | |||
XM_011521837.2 | XP_011520139.1 | 7, 8 | 549 | |||
XM_017022439.1 | XP_016877928.1 | 9, 10 | 780 | |||
XM_017022440.1 | XP_016877929.1 | 8, 9 | 679 | |||
XM_017022441.1 | XP_016877930.1 | 7, 8 | 998 | |||
GenBank mRNA | AK122666.1 | - | 7, 8 | 781 | ||
CR627426.1 | - | 9, 10 | 767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
