siRNA Details
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020759.2 | NP_065810.2 | 13, 14 | 1230 | ||
XM_011521831.2 | XP_011520133.1 | 13, 14 | 1243 | |||
XM_011521832.2 | XP_011520134.1 | 13, 14 | 1240 | |||
XM_011521833.2 | XP_011520135.1 | 12, 13 | 1210 | |||
XM_011521834.2 | XP_011520136.1 | 13, 14 | 1243 | |||
XM_011521836.2 | XP_011520138.1 | 13, 14 | 1545 | |||
XM_011521837.2 | XP_011520139.1 | 11, 12 | 1012 | |||
XM_017022440.1 | XP_016877929.1 | 12, 13 | 1142 | |||
XM_017022441.1 | XP_016877930.1 | 11, 12 | 1461 | |||
XM_017022442.1 | XP_016877931.1 | 6, 7 | 569 | |||
XM_017022443.1 | XP_016877932.1 | 5, 6 | 553 | |||
GenBank mRNA | CR627426.1 | - | 11, 12 | 1948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
