siRNA Details
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020759.2 | NP_065810.2 | 19 | 1781 | ||
XM_011521831.2 | XP_011520133.1 | 19 | 1794 | |||
XM_011521832.2 | XP_011520134.1 | 19 | 1791 | |||
XM_011521833.2 | XP_011520135.1 | 18 | 1761 | |||
XM_011521834.2 | XP_011520136.1 | 18 | 1731 | |||
XM_011521835.2 | XP_011520137.1 | 17 | 1653 | |||
XM_011521836.2 | XP_011520138.1 | 19 | 2096 | |||
XM_011521837.2 | XP_011520139.1 | 17 | 1563 | |||
XM_011521839.2 | XP_011520141.1 | 6 | 563 | |||
XM_017022439.1 | XP_016877928.1 | 18 | 1716 | |||
XM_017022440.1 | XP_016877929.1 | 18 | 1693 | |||
XM_017022441.1 | XP_016877930.1 | 17 | 2012 | |||
XM_017022442.1 | XP_016877931.1 | 12 | 1120 | |||
XM_017022443.1 | XP_016877932.1 | 11 | 1104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
