siRNA Details
Gene Symbol: LINC00308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038400.1 | 4 | 328 | |||
GenBank mRNA | AY077696.1 | - | 4 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
