siRNA Details
Gene Symbol: PGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042371.2 | NP_001035830.1 | 2 | 934 | ||
GenBank mRNA | BC035985.1 | - | 2 | 898 | ||
DA336356.1 | - | 1 | 30 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
