siRNA Details
Gene Symbol: MTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000254.2 | NP_000245.2 | 33 | 8153 | ||
NM_001291939.1 | NP_001278868.1 | 32 | 8000 | |||
NM_001291940.1 | NP_001278869.1 | 32 | 8040 | |||
XM_005273141.4 | XP_005273198.1 | 33 | 8122 | |||
XM_006711770.2 | XP_006711833.1 | 27 | 7101 | |||
XM_011544194.2 | XP_011542496.1 | 32 | 8915 | |||
XM_017001329.1 | XP_016856818.1 | 31 | 8762 | |||
XM_017001330.1 | XP_016856819.1 | 30 | 8726 | |||
GenBank mRNA | AK090487.1 | - | 1 | 22 | ||
AK125206.1 | - | 1 | 657 | |||
BC015894.1 | - | 1 | 609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
