siRNA Details
Gene Symbol: CNTNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201380.2 | NP_001188309.2 | 24 | 5966 | ||
GenBank mRNA | AK094831.1 | - | 1 | 77 | ||
BX538190.1 | - | 5 | 2234 |
Gene Symbol: CNTNAP3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111893.2 | 24 | 5655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
