siRNA Details
Gene Symbol: LOC643802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207030.1 | NP_001193959.1 | 2 | 707 | ||
GenBank mRNA | BX537874.1 | - | 2 | 548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
