siRNA Details
Gene Symbol: PATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145112.1 | NP_001138584.1 | 8 | 793 | ||
XM_011521336.2 | XP_011519638.2 | 8 | 1619 | |||
XM_011521337.2 | XP_011519639.2 | 8 | 1754 | |||
XM_011521338.2 | XP_011519640.1 | 10 | 1318 | |||
XM_011521339.2 | XP_011519641.1 | 9 | 1199 | |||
XM_011521340.2 | XP_011519642.1 | 8 | 1140 | |||
XM_011521341.1 | XP_011519643.1 | 8 | 903 | |||
XM_011521342.2 | XP_011519644.1 | 6 | 1800 | |||
XM_011521343.2 | XP_011519645.1 | 9 | 1150 | |||
XM_011521344.2 | XP_011519646.1 | 6 | 592 | |||
XM_011521345.2 | XP_011519647.1 | 9 | 1112 | |||
XM_011521346.2 | XP_011519648.2 | 4 | 1326 | |||
XM_011521347.1 | XP_011519649.1 | 7 | 708 | |||
XM_011521348.2 | XP_011519650.1 | 7 | 708 | |||
XM_017022000.1 | XP_016877489.1 | 8 | 1611 | |||
XM_017022001.1 | XP_016877490.1 | 6 | 740 | |||
GenBank mRNA | BI835136.1 | - | 7 | 708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
