siRNA Details
Gene Symbol: PATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145112.1 | NP_001138584.1 | 14 | 1546 | ||
XM_011521348.2 | XP_011519650.1 | 13 | 1461 | |||
XM_017022000.1 | XP_016877489.1 | 14 | 2364 | |||
GenBank mRNA | BI909158.1 | - | 6 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
