siRNA Details
Gene Symbol: BEX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012978.2 | NP_001012996.1 | 3 | 423 | ||
NM_001159560.1 | NP_001153032.1 | 3 | 465 | |||
GenBank mRNA | AY833564.1 | - | 3 | 371 | ||
BC042818.1 | - | 3 | 372 | 5313992 | ||
BC092450.1 | - | 3 | 343 | 30720892 | ||
BC106955.2 | - | 3 | 424 | 40035908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
