siRNA Details
Gene Symbol: C12orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099676.2 | NP_001093146.1 | 3 | 472 | ||
NM_001170633.1 | NP_001164104.1 | 3 | 472 | |||
XM_017018770.1 | XP_016874259.1 | 3 | 487 | |||
GenBank mRNA | BC039369.1 | - | 3 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
