siRNA Details
Gene Symbol: C12orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099676.2 | NP_001093146.1 | 7 | 939 | ||
NM_001170633.1 | NP_001164104.1 | 9 | 1419 | |||
GenBank mRNA | BC015121.1 | - | 4 | 410 | ||
BC039369.1 | - | 7 | 887 | |||
BX089271.1 | - | 4 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
