siRNA Details
Gene Symbol: TUSC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172367.2 | NP_758955.2 | 3 | 2938 | ||
GenBank mRNA | AL831929.1 | - | 4 | 2334 | ||
BC142615.1 | - | 3 | 2922 | 40147346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
