siRNA Details
Gene Symbol: SPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010862.2 | NP_001010862.2 | 2 | 653 | ||
XM_006724577.3 | XP_006724640.1 | 2 | 714 | |||
XM_006724578.3 | XP_006724641.1 | 2 | 621 | |||
XM_017029315.1 | XP_016884804.1 | 2 | 1056 | |||
GenBank mRNA | AK127485.1 | - | 1 | 103 | ||
AK304047.1 | - | 2 | 590 | |||
AL832091.1 | - | 2 | 310 | |||
BC146899.1 | - | 2 | 374 | 9056827 | ||
BC146906.1 | - | 2 | 374 | 9056834 | ||
BX403408.2 | - | 2 | 601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
