siRNA Details
Gene Symbol: GTSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278515.1 | NP_001265444.1 | 2 | 417 | ||
GenBank mRNA | AY262164.1 | - | 2 | 417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
