siRNA Details
Gene Symbol: GSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017439.3 | NP_059135.2 | 11, 12 | 856 | ||
XM_011516325.2 | XP_011514627.1 | 11, 12 | 869 | |||
XM_011516326.2 | XP_011514628.1 | 11, 12 | 869 | |||
XM_011516327.2 | XP_011514629.1 | 11, 12 | 869 | |||
XM_011516330.2 | XP_011514632.1 | 4, 5 | 315 | |||
XM_011516331.2 | XP_011514633.1 | 11, 12 | 869 | |||
XM_011516332.2 | XP_011514634.1 | 11, 12 | 869 | |||
XM_017012343.1 | XP_016867832.1 | 11, 12 | 869 | |||
XM_017012344.1 | XP_016867833.1 | 11, 12 | 877 | |||
XM_017012345.1 | XP_016867834.1 | 11, 12 | 869 | |||
XM_017012346.1 | XP_016867835.1 | 9, 10 | 738 | |||
XM_017012347.1 | XP_016867836.1 | 9, 10 | 751 | |||
XM_017012348.1 | XP_016867837.1 | 12, 13 | 945 | |||
XM_017012349.1 | XP_016867838.1 | 11, 12 | 1316 | |||
XM_017012350.1 | XP_016867839.1 | 11, 12 | 869 | |||
XM_017012351.1 | XP_016867840.1 | 11, 12 | 869 | |||
GenBank mRNA | AL834358.1 | - | 10, 11 | 673 | ||
BC110797.1 | - | 11, 12 | 856 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
