siRNA Details
Gene Symbol: TMEM232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039763.3 | NP_001034852.3 | 7 | 829 | ||
XM_006714670.3 | XP_006714733.1 | 9 | 1205 | |||
XM_011543552.2 | XP_011541854.1 | 10 | 1556 | |||
XM_011543553.2 | XP_011541855.1 | 7 | 1639 | |||
XM_011543555.2 | XP_011541857.1 | 9 | 1420 | |||
XM_011543556.1 | XP_011541858.1 | 7 | 1150 | |||
XM_011543557.2 | XP_011541859.1 | 10 | 1557 | |||
XM_011543559.2 | XP_011541861.1 | 10 | 1556 | |||
XM_011543560.2 | XP_011541862.1 | 5 | 529 | |||
XM_011543561.2 | XP_011541863.1 | 10 | 1556 | |||
XM_011543563.2 | XP_011541865.1 | 10 | 1555 | |||
XM_011543564.2 | XP_011541866.1 | 10 | 1554 | |||
XM_011543565.2 | XP_011541867.1 | 10 | 1554 | |||
XM_011543566.2 | XP_011541868.1 | 7 | 911 | |||
XM_011543567.2 | XP_011541869.1 | 10 | 1554 | |||
XM_017009704.1 | XP_016865193.1 | 8 | 1785 | |||
XM_017009705.1 | XP_016865194.1 | 10 | 1557 | |||
XM_017009706.1 | XP_016865195.1 | 10 | 1556 | |||
XM_017009707.1 | XP_016865196.1 | 8 | 1048 | |||
GenBank mRNA | AK125070.1 | - | 1 | 257 | ||
AK296542.1 | - | 7 | 745 | |||
BC042563.1 | - | 6 | 612 | |||
BM465182.1 | - | 5 | 534 | |||
BX648527.1 | - | 4 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
