siRNA Details
Gene Symbol: MATN1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034182.1 | 4 | 1825 | |||
GenBank mRNA | AK057725.1 | - | 4 | 1825 | ||
AK128076.1 | - | 5 | 4058 | |||
BC044253.1 | - | 4 | 1572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
