siRNA Details
Gene Symbol: TEX38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145474.3 | NP_001138946.1 | 2 | 303 | ||
NM_001300863.1 | NP_001287792.1 | 2 | 224 | |||
NM_001300864.1 | NP_001287793.1 | 2 | 152 | |||
XM_011541421.2 | XP_011539723.1 | 2 | 299 | |||
GenBank mRNA | BC057818.1 | - | 2 | 282 | ||
BC067853.1 | - | 2 | 308 | |||
BC126318.1 | - | 2 | 258 | |||
BC144026.1 | - | 2 | 179 | 9052546 | ||
BC144027.1 | - | 2 | 107 | 9052547 | ||
FJ211596.1 | - | 2 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
