siRNA Details
Gene Symbol: TEX38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145474.3 | NP_001138946.1 | 2 | 193 | ||
XM_011541421.2 | XP_011539723.1 | 2 | 189 | |||
GenBank mRNA | BC057818.1 | - | 2 | 172 | ||
BC067853.1 | - | 2 | 198 | |||
BC126318.1 | - | 2 | 148 | |||
FJ211596.1 | - | 2 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
