siRNA Details
Gene Symbol: ZNF749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023561.3 | NP_001018855.2 | 3 | 1140 | ||
NM_001321952.1 | NP_001308881.1 | 3 | 1214 | |||
NM_001321953.1 | NP_001308882.1 | 2 | 1087 | |||
NM_001321954.1 | NP_001308883.1 | 2 | 1013 | |||
XM_011526956.2 | XP_011525258.1 | 3 | 889 | |||
XM_017026802.1 | XP_016882291.1 | 4 | 2140 | |||
GenBank mRNA | AK122794.1 | - | 3 | 1136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
