siRNA Details
Gene Symbol: BMP8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181809.3 | NP_861525.2 | 7 | 4362 | ||
GenBank mRNA | AK024413.1 | - | 1 | 775 | ||
AK127199.1 | - | 1 | 1947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
