siRNA Details
Gene Symbol: TBX5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038440.1 | 3 | 2661 | |||
GenBank mRNA | AK091766.1 | - | 2 | 2644 | ||
U47671.1 | - | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
