siRNA Details
Gene Symbol: LINC00221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027457.2 | 3 | 931 | |||
GenBank mRNA | BX161407.1 | - | 2 | 734 | ||
BX248748.1 | - | 3 | 903 | |||
BX356743.2 | - | 3 | 857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
