siRNA Details
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145290.3 | NP_660333.2 | 4, 5 | 739 | ||
XM_005248137.4 | XP_005248194.1 | 4, 5 | 743 | |||
XM_017007834.1 | XP_016863323.1 | 4, 5 | 265 | |||
GenBank mRNA | AK299168.1 | - | 1, 2 | 26 | ||
AY181243.1 | - | 2, 3 | 94 | |||
AY358306.1 | - | 4, 5 | 682 | |||
BC035645.1 | - | 4, 5 | 750 | |||
BX416923.2 | - | 4, 5 | 729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
