siRNA Details
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145290.3 | NP_660333.2 | 6 | 913 | ||
XM_005248137.4 | XP_005248194.1 | 6 | 917 | |||
XM_017007834.1 | XP_016863323.1 | 6 | 439 | |||
GenBank mRNA | AK095866.1 | - | 1 | 95 | ||
AK299168.1 | - | 3 | 200 | |||
AY181243.1 | - | 4 | 268 | |||
AY358306.1 | - | 6 | 856 | |||
BC035645.1 | - | 6 | 924 | |||
BX416923.2 | - | 6 | 903 |
Gene Symbol: LOC100996333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL359580.2 | - | 1 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
