siRNA Details
Gene Symbol: SOWAHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029870.1 | NP_001025041.1 | 1 | 218 | ||
GenBank mRNA | BC137241.1 | - | 1 | 295 | 9021238 | |
BC137255.1 | - | 1 | 295 | 9021252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
